Merck
  • Detection of microRNAs by the combination of Exonuclease-III assisted target recycling amplification and repeated-fishing strategy.
Detection of microRNAs by the combination of Exonuclease-III assisted target recycling amplification and repeated-fishing strategy.
Analytica chimica acta (2020-09-16)
Xiao-Mei Yan, Yin-Qi Wang, Yao Chen, Zeng-Ping Chen, Ru-Qin Yu
ZUSAMMENFASSUNG
A simple but effective method for the detection of miRNAs was proposed by integrating exonuclease-III assisted target recycling amplification and repeated-fishing strategy. In the proposed method, exonuclease-III assisted target recycling amplification reaction is adopted to produce a large amount of DNA fragments with fluorescence group at its 5' end in the presence of the target miRNA, which are then repeatedly fished out from the reaction mixture by a gold foil modified with a capture probe and transferred into a so-called 'product tube'. The amount of the target miRNA can then be determined from the fluorescence measurement of the solution in the 'product tube'. Application to the detection of miRNA-155 in samples of KH-2 and BRSA-2B cells revealed that the proposed method could achieve sensitive and accurate quantification of the target miRNA with a limit of detection of 36 fM and recovery rates in the range from 96.2% to 105%. Its simplicity, sensitivity and resistance to possible fluorescence interferences in complex biological samples make the proposed method a potentially competitive alternative for miRNAs detection in complex biological samples.