Merck
  • 1-Bromo-3-(1',1'-dimethylalkyl)-1-deoxy-Δ(8)-tetrahydrocannabinols: New selective ligands for the cannabinoid CB(2) receptor.
1-Bromo-3-(1',1'-dimethylalkyl)-1-deoxy-Δ(8)-tetrahydrocannabinols: New selective ligands for the cannabinoid CB(2) receptor.
Bioorganic & medicinal chemistry (2010-10-15)
John W Huffman, Seon A Hepburn, Nataliya Lyutenko, Alicia L S Thompson, Jenny L Wiley, Dana E Selley, Billy R Martin
ZUSAMMENFASSUNG
Δ(8)-Tetrahydrocannabinol (26), 3-(1',1'-dimethylbutyl)- (12), 3-(1',1'-dimethylpentyl)- (13), 3-(1',1'-dimethylhexyl)- (14) and 3-(1',1'-dimethylheptyl)-Δ(8)-tetrahydrocannabinol (15) have been converted into the corresponding 1-bromo-1-deoxy-Δ(8)-tetrahydrocannabinols (25, 8-11). This was accomplished using a protocol developed in our laboratory in which the trifluoromethanesulfonate of a phenol undergoes palladium mediated coupling with pinacolborane. Reaction of this dioxaborolane with aqueous-methanolic copper(II) bromide provides the aryl bromide. The affinities of these bromo cannabinoids for the cannabinoid CB(1) and CB(2) receptors were determined. All of these compounds showed selectivity for the CB(2) receptor and one of them, 1-bromo-1-deoxy-3-(1',1'-dimethylhexyl)-Δ(8)-tetrahydrocannabinol (10), exhibits 52-fold selectivity for this receptor with good (28nM) affinity.