Merck
  • Designed Ankyrin Repeat Protein (DARPin) to target chimeric antigen receptor (CAR)-redirected T cells towards CD4+ T cells to reduce the latent HIV+ cell reservoir.
Designed Ankyrin Repeat Protein (DARPin) to target chimeric antigen receptor (CAR)-redirected T cells towards CD4+ T cells to reduce the latent HIV+ cell reservoir.
Medical microbiology and immunology (2020-09-13)
Lea Patasic, Janna Seifried, Valerie Bezler, Marcell Kaljanac, Irene C Schneider, Heike Schmitz, Christiane Tondera, Jessica Hartmann, Andreas Hombach, Christian J Buchholz, Hinrich Abken, Renate König, Klaus Cichutek
ZUSAMMENFASSUNG
Chimeric Antigen Receptor (CAR)-redirected T cells show great efficacy in the patient-specific therapy of hematologic malignancies. Here, we demonstrate that a DARPin with specificity for CD4 specifically redirects and triggers the activation of CAR engineered T cells resulting in the depletion of CD4+ target cells aiming for elimination of the human immunodeficiency virus (HIV) reservoir.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
SAHA, ≥98% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
1,2-Dilinoleoyl-3-palmitoyl-rac-Glycerin, ≥95% (TLC), liquid
Preise und Verfügbarkeit sind derzeit nicht verfügbar.