Merck
  • Lipoic acid decreases breast cancer cell proliferation by inhibiting IGF-1R via furin downregulation.
Lipoic acid decreases breast cancer cell proliferation by inhibiting IGF-1R via furin downregulation.
British journal of cancer (2020-01-29)
Diana Farhat, Sophie Léon, Sandra E Ghayad, Nicolas Gadot, Philippe Icard, Muriel Le Romancer, Nader Hussein, Hubert Lincet
ZUSAMMENFASSUNG
Breast cancer is the second most common cancer in the world. Despite advances in therapies, the mechanisms of resistance remain the underlying cause of morbidity and mortality. Lipoic acid (LA) is an antioxidant and essential cofactor in oxidative metabolism. Its potential therapeutic effects have been well documented, but its mechanisms of action (MOA) are not fully understood. The aim of this study is to validate the inhibitory LA effect on the proliferation of various breast cancer cell lines and to investigate the MOA that may be involved in this process. We tested LA effects by ex vivo studies on fresh human mammary tumour samples. We demonstrate that LA inhibits the proliferation and Akt and ERK signalling pathways of several breast cancer cells. While searching for upstream dysregulations, we discovered the loss of expression of IGF-1R upon exposure to LA. This decrease is due to the downregulation of the convertase, furin, which is implicated in the maturation of IGF-1R. Moreover, ex vivo studies on human tumour samples showed that LA significantly decreases the expression of the proliferation marker Ki67. LA exerts its anti-proliferative effect by inhibiting the maturation of IGF-1R via the downregulation of furin.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Furin
Preise und Verfügbarkeit sind derzeit nicht verfügbar.