Merck
Inoculum microbiome composition impacts fatty acid product profile from cellulosic feedstock.
Bioresource technology (2021-01-11)
Jorge L Rico, Kenneth F Reardon, Susan K De Long
ZUSAMMENFASSUNG
Conversion of organic wastes to fatty acids rather than methane through anaerobic digestion-based technologies has considerable promise. However, the relationships between microbiome structure and fatty acids produced from cellulosic feedstocks are not well understood. This study investigated the nature of those relationships for anaerobic digester sludge, bison rumen, and cattle rumen inocula grown on cellulose. Acetic acid production was highest in anaerobic sludge reactors, while propionic acid production was highest in cattle rumen reactors. Butyric and pentanoic acid were produced at the highest rates in bison rumen before Day 5. Reactor microbiomes remained distinct, despite identical operating conditions. Novel associations linked Alistipes with butyric acid production and Eubacterium nodatum and Clostridiales bacterium with pentanoic acid production. This study provides new insights into the ability of microbiomes to convert cellulose to different fatty acid mixtures and adds impetus for the rewiring of anaerobic digestion to generate high-value products.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Iodoform, 99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.