Merck
  • Metabolomics investigation of summer mortality in New Zealand Greenshell™ mussels (Perna canaliculus).
Metabolomics investigation of summer mortality in New Zealand Greenshell™ mussels (Perna canaliculus).
Fish & shellfish immunology (2020-08-17)
Thao V Nguyen, Andrea C Alfaro
ZUSAMMENFASSUNG
Increasing water temperatures due to climate change have resulted in more frequent high mortality events of New Zealand Greenshell™ mussels (Perna canaliculus Gmelin 1791). These events have significant impacts within mussel farms which support a major shellfish industry for New Zealand. The present study investigates metabolic responses of farmed mussels during a summer mortality event in order to identify health impacts and elucidate mechanistic effects of external stressors on mussels. A gas chromatography-mass spectrometry (GC-MS)-based metabolomics approach was used to identify metabolic perturbations and flow cytometry assays were used to assess viability, oxidative stress and apoptosis of haemocytes from healthy and unhealthy mussels during a summer mortality event. The results showed significantly higher mortality and apoptosis of haemocytes in unhealthy mussels compared to healthy mussels. Reactive oxygen species (ROS) production, which is an indicator of oxidative stress was very high in both mussel groups, but no differences were observed between the two mussel groups. Metabolomics revealed alterations of many metabolites in both haemolymph and hepatopancreas (digestive gland) of unhealthy mussels compared to healthy mussels, reflecting perturbations in several molecular pathways, including energy metabolism, amino acid metabolism, protein degradation/tissue damage and oxidative stress. An increased level of itaconic acid which is an antimicrobial metabolite and biomarker of pathogen infection was observed in haemolymph, but not in hepatopancreas samples. This investigation provides the first detailed metabolic characterization of mussel immune responses to a summer mortality event and illustrates the benefits of using an integrated metabolomics and flow cytometry workflow for mussel health assessment and biomarker identification for summer mortality early detection.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methylchlorameisensäureester, 99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.