Merck
Quinacrine-CASIN combination overcomes chemoresistance in human acute lymphoid leukemia.
Nature communications (2021-11-28)
Limei Wu, Srinivas Chatla, Qiqi Lin, Fabliha Ahmed Chowdhury, Werner Geldenhuys, Wei Du
ZUSAMMENFASSUNG
Chemoresistance posts a major hurdle for treatment of acute leukemia. There is increasing evidence that prolonged and intensive chemotherapy often fails to eradicate leukemic stem cells, which are protected by the bone marrow niche and can induce relapse. Thus, new therapeutic approaches to overcome chemoresistance are urgently needed. By conducting an ex vivo small molecule screen, here we have identified Quinacrine (QC) as a sensitizer for Cytarabine (AraC) in treating acute lymphoblastic leukemia (ALL). We show that QC enhances AraC-mediated killing of ALL cells, and subsequently abrogates AraC resistance both in vitro and in an ALL-xenograft model. However, while combo AraC+QC treatment prolongs the survival of primary transplanted recipients, the combination exhibits limited efficacy in secondary transplanted recipients, consistent with the survival of niche-protected leukemia stem cells. Introduction of Cdc42 Activity Specific Inhibitor, CASIN, enhances the eradication of ALL leukemia stem cells by AraC+QC and prolongs the survival of both primary and secondary transplanted recipients without affecting normal long-term human hematopoiesis. Together, our findings identify a small-molecule regimen that sensitizes AraC-mediated leukemia eradication and provide a potential therapeutic approach for better ALL treatment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-74, purified immunoglobulin, buffered aqueous solution
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Cytosin β-D-Arabinofuranosid, crystalline, ≥90% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.