Merck
Protocol for evaluating protein relocalization from the plasma membrane to chloroplasts.
STAR protocols (2021-09-30)
Laura Medina-Puche, Chanhong Kim, Rosa Lozano-Duran, Vivek Dogra
ZUSAMMENFASSUNG
We present a protocol for analyzing and evaluating the relocalization of proteins from the plasma membrane to chloroplasts. Some plant membrane-bound proteins carry dual targeting signals, e.g., a membrane-anchoring N-myristoylation motif and a chloroplast transit peptide. These proteins are predominantly targeted to membranes; upon certain cues, however, they can undergo detachment from membranes and relocalization to chloroplasts. This protocol combines imaging and biochemical analyses to track in a reliable and quantitative manner the relocalization of proteins between subcellular organelles. For complete details on the use and execution of this protocol, please refer to Medina-Puche et al. (2020).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Proteasehemmer-Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
TWEEN® 20, Molecular Biology, viscous liquid
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, BioReagent, suitable for electrophoresis, ≥99.0%
Sigma-Aldrich
Ammoniumpersulfat, Molecular Biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
Kaliumchlorid, Molecular Biology, ≥99.0%
Sigma-Aldrich
Ethylenglykol-bis(2-aminoethylether)-N,N,N′,N′-Tetraessigsäure, Molecular Biology, ≥97.0%
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
4′-Hydroxy-3′,5′-dimethoxyacetophenon, 97%
Sigma-Aldrich
DL-Dithiothreitol, ≥99.0% (RT)
Sigma-Aldrich
Natriumphosphat, BioXtra, ≥99.0%
Sigma-Aldrich
Ficoll® 400, BioXtra, lyophilized powder
Sigma-Aldrich
Natriumorthovanadat, 99.98% trace metals basis
Sigma-Aldrich
Bromphenolblau, titration: suitable