Merck
  • Development and application of 15N-tracer substances for measuring the whole-body protein turnover rates in the human, especially in neonates: a review.
Development and application of 15N-tracer substances for measuring the whole-body protein turnover rates in the human, especially in neonates: a review.
Isotopes in environmental and health studies (2012-03-09)
Klaus D Wutzke
ZUSAMMENFASSUNG
Our research group of the Children's Hospital of the University of Rostock (Rostock group) has long-time experience in (15)N-labelling and in using yeast protein and its hydrolysates for tracer kinetic studies to evaluate parameters of the whole-body protein metabolism in premature infants. The particular advantage of applying an economically convenient, highly (15)N-enriched, and completely labelled yeast protein for evaluating protein turnover rates is the fact that the (15)N dose is spread among all proteinogenic amino acids. The absorption has been improved by hydrolysing [(15)N]yeast protein with thermitase into a mixture of amino acids, dipeptides and tripeptides so that faecal analysis becomes unnecessary when determining turnover rates. The review shows that, in contrast to the application of single (15)N-labelled amino acids with resulting overestimation of protein turnover rates, the (15)N-labelled yeast protein thermitase hydrolysate represents the amino acid metabolism more closely without causing amino acid imbalances. The (15)N-labelled yeast protein thermitase hydrolysate leads to the estimation of reliable protein turnover rates, particularly in premature infants.