Merck
  • SFPQ rescues F508del-CFTR expression and function in cystic fibrosis bronchial epithelial cells.
SFPQ rescues F508del-CFTR expression and function in cystic fibrosis bronchial epithelial cells.
Scientific reports (2021-08-19)
Parameet Kumar, Dharmendra Kumar Soni, Chaitali Sen, Mads B Larsen, Krystyna Mazan-Mamczarz, Yulan Piao, Supriyo De, Myriam Gorospe, Raymond A Frizzell, Roopa Biswas
ZUSAMMENFASSUNG
Cystic fibrosis (CF) occurs as a result of mutations in the cystic fibrosis transmembrane conductance regulator (CFTR) gene, which lead to misfolding, trafficking defects, and impaired function of the CFTR protein. Splicing factor proline/glutamine-rich (SFPQ) is a multifunctional nuclear RNA-binding protein (RBP) implicated in the regulation of gene expression pathways and intracellular trafficking. Here, we investigated the role of SFPQ in the regulation of the expression and function of F508del-CFTR in CF lung epithelial cells. We find that the expression of SFPQ is reduced in F508del-CFTR CF epithelial cells compared to WT-CFTR control cells. Interestingly, the overexpression of SFPQ in CF cells increases the expression as well as rescues the function of F508del-CFTR. Further, comprehensive transcriptome analyses indicate that SFPQ plays a key role in activating the mutant F508del-CFTR by modulating several cellular signaling pathways. This is the first report on the role of SFPQ in the regulation of expression and function of F508del-CFTR in CF lung disease. Our findings provide new insights into SFPQ-mediated molecular mechanisms and point to possible novel epigenetic therapeutic targets for CF and related pulmonary diseases.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phosphatasehemmer-Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
SIGMAFAST Proteaseinhibitor-Tabletten, For General Use
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Anti-Glyceraldehyd-3-phosphat-dehydrogenase-Antikörper, Klon 6C5, clone 6C5, Chemicon®, from mouse
Preise und Verfügbarkeit sind derzeit nicht verfügbar.