Merck
  • A single-cell Arabidopsis root atlas reveals developmental trajectories in wild-type and cell identity mutants.
A single-cell Arabidopsis root atlas reveals developmental trajectories in wild-type and cell identity mutants.
Developmental cell (2022-02-09)
Rachel Shahan, Che-Wei Hsu, Trevor M Nolan, Benjamin J Cole, Isaiah W Taylor, Laura Greenstreet, Stephen Zhang, Anton Afanassiev, Anna Hendrika Cornelia Vlot, Geoffrey Schiebinger, Philip N Benfey, Uwe Ohler
ZUSAMMENFASSUNG
In all multicellular organisms, transcriptional networks orchestrate organ development. The Arabidopsis root, with its simple structure and indeterminate growth, is an ideal model for investigating the spatiotemporal transcriptional signatures underlying developmental trajectories. To map gene expression dynamics across root cell types and developmental time, we built a comprehensive, organ-scale atlas at single-cell resolution. In addition to estimating developmental progressions in pseudotime, we employed the mathematical concept of optimal transport to infer developmental trajectories and identify their underlying regulators. To demonstrate the utility of the atlas to interpret new datasets, we profiled mutants for two key transcriptional regulators at single-cell resolution, shortroot and scarecrow. We report transcriptomic and in vivo evidence for tissue trans-differentiation underlying a mixed cell identity phenotype in scarecrow. Our results support the atlas as a rich community resource for unraveling the transcriptional programs that specify and maintain cell identity to regulate spatiotemporal organ development.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, pH 5.2, ≥96%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Pektolyase aus Aspergillus japonicus, lyophilized powder, ≥0.3 units/mg solid
Preise und Verfügbarkeit sind derzeit nicht verfügbar.