Merck
Identification of a 3-Gene Prognostic Index for Papillary Thyroid Carcinoma.
Frontiers in molecular biosciences (2022-04-05)
Lin-Kun Zhong, Xing-Yan Deng, Fei Shen, Wen-Song Cai, Jian-Hua Feng, Xiao-Xiong Gan, Shan Jiang, Chi-Zhuai Liu, Ming-Guang Zhang, Jiang-Wei Deng, Bing-Xing Zheng, Xiao-Zhang Xie, Li-Qing Ning, Hui Huang, Shan-Shan Chen, Jian-Hang Miao, Bo Xu
ZUSAMMENFASSUNG
The accurate determination of the risk of cancer recurrence is a critical unmet need in managing thyroid cancer (TC). Although numerous studies have successfully demonstrated the use of high throughput molecular diagnostics in TC prediction, it has not been successfully applied in routine clinical use, particularly in Chinese patients. In our study, we objective to screen for characteristic genes specific to PTC and establish an accurate model for diagnosis and prognostic evaluation of PTC. We screen the differentially expressed genes by Python 3.6 in The Cancer Genome Atlas (TCGA) database. We discovered a three-gene signature Gap junction protein beta 4 (GJB4), Ripply transcriptional repressor 3 (RIPPLY3), and Adrenoceptor alpha 1B (ADRA1B) that had a statistically significant difference. Then we used Gene Expression Omnibus (GEO) database to establish a diagnostic and prognostic model to verify the three-gene signature. For experimental validation, immunohistochemistry in tissue microarrays showed that thyroid samples' proteins expressed by this three-gene are differentially expressed. Our protocol discovered a robust three-gene signature that can distinguish prognosis, which will have daily clinical application.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-RIPPLY3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Preise und Verfügbarkeit sind derzeit nicht verfügbar.