Merck
Targeting EP2 receptor with multifaceted mechanisms for high-risk neuroblastoma.
Cell reports (2022-06-23)
Ruida Hou, Ying Yu, Madison N Sluter, Lexiao Li, Jiukuan Hao, Jie Fang, Jun Yang, Jianxiong Jiang
ZUSAMMENFASSUNG
Prostaglandin E2 (PGE2) promotes tumor cell proliferation, migration, and invasion, fostering an inflammation-enriched microenvironment that facilitates angiogenesis and immune evasion. However, the PGE2 receptor subtype (EP1-EP4) involved in neuroblastoma (NB) growth remains elusive. Herein, we show that the EP2 receptor highly correlates with NB aggressiveness and acts as a predominant Gαs-coupled receptor mediating PGE2-initiated cyclic AMP (cAMP) signaling in NB cells with high-risk factors, including 11q deletion and MYCN amplification. Knockout of EP2 in NB cells blocks the development of xenografts, and its conditional knockdown prevents established tumors from progressing. Pharmacological inhibition of EP2 by our recently developed antagonist TG6-129 suppresses the growth of NB xenografts in nude mice and syngeneic allografts in immunocompetent hosts, accompanied by anti-inflammatory, antiangiogenic, and apoptotic effects. This proof-of-concept study suggests that the PGE2/EP2 signaling pathway contributes to NB malignancy and that EP2 inhibition by our drug-like compounds provides a promising strategy to treat this deadly pediatric cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Rolipram, solid, ≥98% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.