Merck
  • The EMT transcription factor ZEB1 governs a fitness-promoting but vulnerable DNA replication stress response.
The EMT transcription factor ZEB1 governs a fitness-promoting but vulnerable DNA replication stress response.
Cell reports (2022-12-15)
Harald Schuhwerk, Julia Kleemann, Pooja Gupta, Ruthger van Roey, Isabell Armstark, Martina Kreileder, Nora Feldker, Vignesh Ramesh, Yussuf Hajjaj, Kathrin Fuchs, Mousumi Mahapatro, Mojca Hribersek, Marco Volante, Arwin Groenewoud, Felix B Engel, Paolo Ceppi, Markus Eckstein, Arndt Hartmann, Fabian Müller, Torsten Kroll, Marc P Stemmler, Simone Brabletz, Thomas Brabletz
ZUSAMMENFASSUNG
The DNA damage response (DDR) and epithelial-to-mesenchymal transition (EMT) are two crucial cellular programs in cancer biology. While the DDR orchestrates cell-cycle progression, DNA repair, and cell death, EMT promotes invasiveness, cellular plasticity, and intratumor heterogeneity. Therapeutic targeting of EMT transcription factors, such as ZEB1, remains challenging, but tumor-promoting DDR alterations elicit specific vulnerabilities. Using multi-omics, inhibitors, and high-content microscopy, we discover a chemoresistant ZEB1-high-expressing sub-population (ZEB1hi) with co-rewired cell-cycle progression and proficient DDR across tumor entities. ZEB1 stimulates accelerated S-phase entry via CDK6, inflicting endogenous DNA replication stress. However, DDR buildups involving constitutive MRE11-dependent fork resection allow homeostatic cycling and enrichment of ZEB1hi cells during transforming growth factor β (TGF-β)-induced EMT and chemotherapy. Thus, ZEB1 promotes G1/S transition to launch a progressive DDR benefitting stress tolerance, which concurrently manifests a targetable vulnerability in chemoresistant ZEB1hi cells. Our study thus highlights the translationally relevant intercept of the DDR and EMT.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Insulin -Lösung human, sterile-filtered, BioXtra, suitable for cell culture
Roche
cOmplete, EDTA-freier Proteasehemmer-Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Puromycin -dihydrochlorid aus Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Choleratoxin aus Vibrio cholerae, ≥90% (SDS-PAGE), lyophilized powder
Sigma-Aldrich
Hydrocortison, BioReagent, suitable for cell culture