Merck
Immunoglobulin-like transcript 2 blockade restores antitumor immune responses in glioblastoma.
Cancer science (2022-09-10)
Seila Lorenzo-Herrero, Christian Sordo-Bahamonde, Alejandra Martínez-Pérez, Mª Daniela Corte-Torres, Iván Fernández-Vega, Mª Pilar Solís-Hernández, Segundo González
ZUSAMMENFASSUNG
Glioblastoma stands as the most frequent primary brain tumor. Despite the multimodal therapy for glioblastoma patients, the survival rate is very low, highlighting the need for novel therapies that improve patient outcomes. Immune checkpoint blockade strategies are achieving promising results in a myriad of tumors and several studies have reported its efficacy in glioblastoma at a preclinical level. ILT2 is a novel immune checkpoint that exerts an inhibitory effect via the interaction with classical and non-classical HLA class-I molecules. Herein, we report that ILT2 blockade promotes antitumor responses against glioblastoma. In silico and immunohistochemical analyses revealed that the expression of ILT2 and its ligands HLA-A, -B, -C, and -E are highly expressed in patients with glioblastoma. Disruption of ILT2 with blocking monoclonal antibodies increased natural killer cell-mediated IFN-γ production and cytotoxicity against glioblastoma, partially reverting the immunosuppression linked to this malignancy. In addition, co-treatment with temozolomide strengthened the antitumor capacity of anti-ILT2-treated immune cells. Collectively, our results establish the basis for future studies regarding the clinical potential of ILT2 blockade alone or in combination regimens in glioblastoma.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-HLA-E antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Preise und Verfügbarkeit sind derzeit nicht verfügbar.