Merck
  • Nicotine, THC, and Dolutegravir Modulate E-Cigarette-Induced Changes in Addiction- and Inflammation-Associated Genes in Rat Brains and Astrocytes.
Nicotine, THC, and Dolutegravir Modulate E-Cigarette-Induced Changes in Addiction- and Inflammation-Associated Genes in Rat Brains and Astrocytes.
Brain sciences (2023-11-25)
Jacqueline Renee Kulbe, Lauren Nguyen, Alexandra Anh Le, Anna Elizabeth Laird, Michael A Taffe, Jacques D Nguyen, Jerel Adam Fields
ZUSAMMENFASSUNG
E-cigarette use has been marketed as a safer alternative to traditional cigarettes, as a means of smoking cessation, and are used at a higher rate than the general population in people with HIV (PWH). Early growth receptor 2 (EGR2) and Activity-Regulated Cytoskeleton-Associated Protein (ARC) have a role in addiction, synaptic plasticity, inflammation, and neurodegeneration. This study showed that 10 days of exposure to e-cigarette vapor altered gene expression in the brains of 6-month-old, male, Sprague Dawley rats. Specifically, the e-cigarette solvent vapor propylene glycol (PG) downregulated EGR2 and ARC mRNA expression in frontal cortex, an effect which was reversed by nicotine (NIC) and THC, suggesting that PG could have a protective role against NIC and cannabis dependence. However, in vitro, PG upregulated EGR2 and ARC mRNA expression at 18 h in cultured C6 rat astrocytes suggesting that PG may have neuroinflammatory effects. PG-induced upregulation of EGR2 and ARC mRNA was reversed by NIC but not THC. The HIV antiretroviral DTG reversed the effect NIC had on decreasing PG-induced upregulation of EGR2, which is concerning because EGR2 has been implicated in HIV latency reversal, T-cell apoptosis, and neuroinflammation, a process that underlies the development of HIV-associated neurocognitive disorders.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-15, ascites fluid
Preise und Verfügbarkeit sind derzeit nicht verfügbar.