Merck
  • Deep Annotation of Donated Chemical Probes (DCP) in Organotypic Human Liver Cultures and Patient-Derived Organoids from Tumor and Normal Colorectum.
Deep Annotation of Donated Chemical Probes (DCP) in Organotypic Human Liver Cultures and Patient-Derived Organoids from Tumor and Normal Colorectum.
ACS chemical biology (2023-03-22)
Claudia Tredup, Benardina Ndreshkjana, Natalie S Schneider, Amelie Tjaden, Aurino M Kemas, Sonia Youhanna, Volker M Lauschke, Benedict-Tilman Berger, Andreas Krämer, Lena M Berger, Sandra Röhm, Stefan Knapp, Henner F Farin, Susanne Müller
ZUSAMMENFASSUNG
Well-characterized small molecules are essential tools for studying the biology and therapeutic relevance of a target protein. However, many compounds reported in the literature and routinely studied in biomedical research lack the potency and selectivity required for mechanistic cellular studies on the function of a given protein. Furthermore, commercially available compounds often do not include useful tools developed by industry as part of their research and development efforts, as they frequently remain proprietary. The freely available donated chemical probe (DCP) library, fueled by generous donations of compounds from industry and academia, enables easy access to a steadily growing collection of these valuable and well-characterized tools. Here, we provide a systematic description of the current DCP library collection and their associated comprehensive characterization data, including a variety of in vitro and cellular assays. Of note, we characterized the set in relevant human primary models by employing hepatotoxicity screening in primary human liver spheroids and viability screening in patient-derived colorectal cancer organoids and matched normal-adjacent epithelium. Taken together, the DCP library represents a well-annotated, openly available collection of tool compounds for studying a wide range of targets, including kinases, G-protein-coupled receptors, and ion channels. As such, it represents a unique resource for the biomedical research community.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
BAY-985, ≥98% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.