Merck
FANCJ DNA helicase is recruited to the replisome by AND-1 to ensure genome stability.
EMBO reports (2024-01-05)
Ana Boavida, Luisa Mr Napolitano, Diana Santos, Giuseppe Cortone, Nanda K Jegadesan, Silvia Onesti, Dana Branzei, Francesca M Pisani
ZUSAMMENFASSUNG
FANCJ, a DNA helicase linked to Fanconi anemia and frequently mutated in cancers, counteracts replication stress by dismantling unconventional DNA secondary structures (such as G-quadruplexes) that occur at the DNA replication fork in certain sequence contexts. However, how FANCJ is recruited to the replisome is unknown. Here, we report that FANCJ directly binds to AND-1 (the vertebrate ortholog of budding yeast Ctf4), a homo-trimeric protein adaptor that connects the CDC45/MCM2-7/GINS replicative DNA helicase with DNA polymerase α and several other factors at DNA replication forks. The interaction between FANCJ and AND-1 requires the integrity of an evolutionarily conserved Ctf4-interacting protein (CIP) box located between the FANCJ helicase motifs IV and V. Disruption of the CIP box significantly reduces FANCJ association with the replisome, causing enhanced DNA damage, decreased replication fork recovery and fork asymmetry in cells unchallenged or treated with Pyridostatin, a G-quadruplex-binder, or Mitomycin C, a DNA inter-strand cross-linking agent. Cancer-relevant FANCJ CIP box variants display reduced AND-1-binding and enhanced DNA damage, a finding that suggests their potential role in cancer predisposition.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonale ANTI-FLAG® M2-Peroxidase (HRP) in Maus hergestellte Antikörper, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Monoklonaler Anti-Polyhistidin−Peroxidase-Antikörper der Maus in Maus hergestellte Antikörper, clone HIS-1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-BACH1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution