Merck
Alterations of epinephrine-induced gluconeogenesis in aging.
Experimental & molecular medicine (2009-03-25)
Kyungtae Kim, Sung Chun Cho, Anthony Cova, Ik Soon Jang, Sang Chul Park
ZUSAMMENFASSUNG
The effects of glucagon and epinephrine on gluconeogenesis in young (4 month) and old (24 month) Fisher 344 rat hepatocytes were compared. In contrast to glucagon, which had a similar effect on gluconeogenesis in both young and old cells, epinephrine caused a smaller increase in gluconeogenesis in old rat hepatocytes than in young hepatocytes. beta2 adrenergic receptor (beta2-AR) expression slightly decreased in aged rat liver, and there were differences between young and old hepatocytes in their patterns of G protein coupled receptor kinases, which are involved in the activation of beta2-AR receptor signal desensitization. The major isoform of the kinase changed from GRK2 to GRK3 and the expression of beta-arrestin, which is recruited by the phosphorylated beta2-AR for internalization and degradation, increased in aged rat liver. GRK3 overexpression also decreased the glucose output from young rat hepatocytes. We conclude that an age-associated reduction in epinephrine-induced gluconeogenesis occurs through the epinephrine receptor desensitizing system.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumpyruvat, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumpyruvat, ReagentPlus®, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Williams‘ Medium E, With sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumpyruvat, powder, BioXtra, suitable for mouse embryo cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Williams‘ Medium E, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumpyruvat, Hybri-Max, powder, suitable for hybridoma
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Williams‘ Medium E, With L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumpyruvat, BioXtra, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.