Merck
  • Mousepox detected in a research facility: case report and failure of mouse antibody production testing to identify Ectromelia virus in contaminated mouse serum.
Mousepox detected in a research facility: case report and failure of mouse antibody production testing to identify Ectromelia virus in contaminated mouse serum.
Comparative medicine (2009-04-25)
Philippe Labelle, Nina E Hahn, Jenelle K Fraser, Lonnie V Kendall, Melanie Ziman, Edward James, Nilabh Shastri, Stephen M Griffey
ZUSAMMENFASSUNG
An outbreak of mousepox in a research institution was caused by Ectromelia-contaminated mouse serum that had been used for bone marrow cell culture and the cells subsequently injected into the footpads of mice. The disease initially was diagnosed by identification of gross and microscopic lesions typical for Ectromelia infection, including foci of necrosis in the liver and spleen and eosinophilic intracytoplasmic inclusion bodies in the skin. The source of infection was determined by PCR analysis to be serum obtained from a commercial vendor. To determine whether viral growth in tissue culture was required to induce viral infection, 36 mice (BALB/cJ, C57BL/6J) were experimentally exposed intraperitoneally, intradermally (footpad), or intranasally to contaminated serum or bone marrow cell cultures using the contaminated serum in the culture medium. Mice were euthanized when clinical signs developed or after 12 wk. Necropsy, PCR of spleen, and serum ELISA were performed on all mice. Mice injected with cell cultures and their cage contacts developed mousepox, antibodies to Ectromelia, and lesions, whereas mice injected with serum without cells did not. Mouse antibody production, a tool commonly used to screen biologic materials for viral contamination, failed to detect active Ectromelia contamination in mouse serum.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Agarose, BioReagent, Molecular Biology, low EEO
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Agarose, Type I, low EEO
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Agarose, Low EEO
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Agarose, Low EEO, for Immunoelectrophoresis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature, molecular biology grade
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Agarose, High EEO
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Agarose, Molecular Biology
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Agarose, Type I-A, low EEO
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Agarose, Medium EEO, Molecular Biology
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Agarose, Type II-A, Medium EEO
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Agarose, For pulsed field electrophoresis running gel
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Agarose, Special High EEO
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Agarose, High Gelling Temperature
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Agarose, Wide range, Molecular Biology
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Agarose, Type IV, Special High EEO
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Agarose, High EEO
Preise und Verfügbarkeit sind derzeit nicht verfügbar.