Merck
Deoxyribose 5-phosphate aldolase of Bacillus cereus: purification and properties.
Biochimica et biophysica acta (1992-01-09)
F Sgarrella, A Del Corso, M G Tozzi, M Camici
ZUSAMMENFASSUNG
Deoxyribose 5-phosphate aldolase was purified 41 times from Bacillus cereus induced by growth on deoxyribonucleosides. The purification procedure includes ammonium sulphate fractionation, gel filtration on Sephadex G-100, ion-exchange chromatography on DEAE-Sephacel and preparative electrophoresis on 10% polyacrylamide gel. The enzyme is stable above pH 6.5, but is rapidly inactivated by sulfhydryl reagents. Being insensitive to EDTA, it may be considered as a Class I aldolase. Among a number of compounds tested (including some carboxylic acids, free and phosphorylated pentoses, nucleotides and nucleosides), none has been found to affect the enzyme activity. The enzyme appears to be dimeric, with a subunit Mr of 23,600. A Km of 4.4 x 10(-4) M was calculated for dRib 5-P.