Merck
  • Isolation and gene expression analysis of a papain-type cysteine protease in thermogenic skunk cabbage (Symplocarpus renifolius).
Isolation and gene expression analysis of a papain-type cysteine protease in thermogenic skunk cabbage (Symplocarpus renifolius).
Bioscience, biotechnology, and biochemistry (2012-10-11)
Yasuko Ito-Inaba, Hiromi Masuko, Masao Watanabe, Takehito Inaba
ZUSAMMENFASSUNG
Skunk cabbage (Symplocarpus renifolius) spadices contain abundant transcripts for cysteine protease (CP). From thermogenic spadices, we isolated SrCPA, a highly expressed CP gene that encoded a papain-type CP. SrCPA is structurally similar to other plant CPs, including the senescence-associated CPs found in aroids. The expression of SrCPA increased during floral development, and was observed in all floral tissues except for the stamens.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Papain aus Carica papaya, solution, light brown, ≥10 U/mg protein (~25 mg/ml)
Sigma-Aldrich
Papain aus papaya latex, crude powder, 1.5-10 units/mg solid
Sigma-Aldrich
Papain aus papaya latex, lyophilized powder, aseptically filled