Merck
Cancer metabolism: what we can learn from proteomic analysis by mass spectrometry.
Cancer genomics & proteomics (2012-11-20)
Weidong Zhou, Lance A Liotta, Emanuel F Petricoin
ZUSAMMENFASSUNG
A variety of genomic and proteomic tools have been used to study cancer metabolism and metabolomics in order to understand how cancer cells survive in their environment. Throughout the past decade, mass spectrometry has been routinely used for large-scale protein identification of complex biological mixtures. In this review, we discuss some recent developments in cancer metabolism by proteomic analysis using mass spectrometric techniques, focusing on pyruvate kinase, L-lactate dehydrogenase, Warburg effect, glutamine metabolism and oxidative stress.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyruvat-Kinase aus Kaninchenmuskel, Type III, lyophilized powder, 350-600 units/mg protein
Sigma-Aldrich
Pyruvat-Kinase aus Kaninchenmuskel, Type II, ammonium sulfate suspension, 350-600 units/mg protein
Sigma-Aldrich
Pyruvat-Kinase aus Kaninchenmuskel, Type VII, buffered aqueous glycerol solution, 350-600 units/mg protein