Merck
  • Safety and pharmacodynamic effects of a pharmacological chaperone on α-galactosidase A activity and globotriaosylceramide clearance in Fabry disease: report from two phase 2 clinical studies.
Safety and pharmacodynamic effects of a pharmacological chaperone on α-galactosidase A activity and globotriaosylceramide clearance in Fabry disease: report from two phase 2 clinical studies.
Orphanet journal of rare diseases (2012-11-28)
Dominique P Germain, Roberto Giugliani, Derralynn A Hughes, Atul Mehta, Kathy Nicholls, Laura Barisoni, Charles J Jennette, Alexander Bragat, Jeff Castelli, Sheela Sitaraman, David J Lockhart, Pol F Boudes
ZUSAMMENFASSUNG
Fabry disease (FD) is a genetic disorder resulting from deficiency of the lysosomal enzyme α-galactosidase A (α-Gal A), which leads to globotriaosylceramide (GL-3) accumulation in multiple tissues. We report on the safety and pharmacodynamics of migalastat hydrochloride, an investigational pharmacological chaperone given orally at 150 mg every-other-day. Two open-label uncontrolled phase 2 studies of 12 and 24 weeks (NCT00283959 and NCT00283933) in 9 males with FD were combined. At multiple time points, α-Gal A activity and GL-3 levels were quantified in blood cells, kidney and skin. GL-3 levels were also evaluated through skin and renal histology. Compared to baseline, increased α-Gal A activity of at least 50% was demonstrated in blood, skin and kidney in 6 of 9 patients. Patients' increased α-Gal A activities paralleled the α-Gal A increases observed in vitro in HEK-293 cells transfected with the corresponding mutant form of the enzyme. The same 6 patients who demonstrated increases of α-Gal A activity also had GL-3 reduction in skin, urine and/or kidney, and had α-Gal A mutations that responded in transfected cells incubated with the drug. The 3 patients who did not show a consistent response in vivo had α-Gal A mutations that did not respond to migalastat HCl in transfected cells. Migalastat HCl was well tolerated. Migalastat HCl is a candidate pharmacological chaperone that provides a novel genotype-specific treatment for FD. It enhanced α-Gal A activity and resulted in GL-3 substrate decrease in patients with responsive GLA mutations. Phase 3 studies are ongoing.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
α-Galactosidase aus green coffee beans, ammonium sulfate suspension, ≥9 units/mg protein
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
α-Galactosidase, positionally specific from Escherichia coli, recombinant, expressed in E. coli, buffered aqueous solution
Preise und Verfügbarkeit sind derzeit nicht verfügbar.