Merck
  • Screening of whey protein isolate hydrolysates for their dual functionality: influence of heat pre-treatment and enzyme specificity.
Screening of whey protein isolate hydrolysates for their dual functionality: influence of heat pre-treatment and enzyme specificity.
Food chemistry (2012-12-01)
Randy Adjonu, Gregory Doran, Peter Torley, Samson Agboola
ZUSAMMENFASSUNG
Heat pre-treated and non heat pre-treated whey protein isolate (WPI) were hydrolysed using α-chymotrypsin (chymotrypsin), pepsin and trypsin. The in vitro antioxidant activity, ACE-inhibition activity and surface hydrophobicities of the hydrolysates were measured in order to determine if peptides with dual functionalities were present. Dual functional peptides have both biological (e.g. antioxidant, ACE-inhibition, opioid activities) and technological (e.g. nanoemulsification abilities) functions in food systems. Heat pre-treatment marginally enhanced the hydrolysis of WPI by pepsin and trypsin but had no effect on WPI hydrolysis with chymotrypsin. With the exception of the hydrolysis by trypsin, heat pre-treatment did not affect the peptide profile of the hydrolysates as analysed using size exclusion chromatography, or the antioxidant activity (P>0.05). Heat pre-treatment significantly affected the ACE-inhibition activities and the surface hydrophobicities of the hydrolysates (P<0.05), which was a function of the specificity of the hydrolysing enzyme. Extended hydrolysis (up to 24 h) had no significant effect on the DH and the molecular weight profiles (P>0.05) but in some instances caused a reduction in the antioxidant activity of WPI hydrolysates. The chymotrypsin hydrolysate showed a broad MW size range, and was followed by pepsin and then trypsin. The bioactivities of the hydrolysates generally decreased in the order; chymotrypsin>trypsin>pepsin. This study showed that by manipulating protein conformation with pre-hydrolysis heat treatment, combined with careful enzyme selection, peptides with dual functionalities can be produced from WPI for use as functional ingredients in the manufacture of functional foods.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pepsin aus Schweinemagenschleimhaut, powder, ≥250 units/mg solid
Sigma-Aldrich
Pepsin aus Schweinemagenschleimhaut, lyophilized powder, ≥3,200 units/mg protein
Sigma-Aldrich
αα-Chymotrypsin aus Rinderpankreas, Type II, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
Pepsin aus Schweinemagenschleimhaut, powder, ≥400 units/mg protein
Sigma-Aldrich
αα-Chymotrypsin, (TLCK treated to inactivate residual tryspin activity), Type VII, essentially salt-free, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
Pepsin aus Schweinemagenschleimhaut, powder, slightly beige, ≥500 U/mg
Sigma-Aldrich
αα-Chymotrypsin aus Rinderpankreas, Type I-S, essentially salt-free, lyophilized powder
Sigma-Aldrich
Pepsin aus Schweinemagenschleimhaut, powder, slightly beige, 1200-2400 U/mg
Sigma-Aldrich
αα-Chymotrypsin aus Rinderpankreas, ≥40 units/mg protein, vial of 5 mg
Sigma-Aldrich
Pepsin aus Schweinemagenschleimhaut, tested according to Ph. Eur.
Sigma-Aldrich
Pepsin−Agarose from porcine gastric mucosa, lyophilized powder, ≥30 units/mg dry solid
Sigma-Aldrich
αα-Chymotrypsin aus Rinderpankreas, suitable for protein sequencing, salt-free, lyophilized powder
Sigma-Aldrich
α-Chymotrypsin from human pancreas, lyophilized powder
Sigma-Aldrich
α-Chymotrypsin−Agarose from bovine pancreas, lyophilized powder, 2,000-3,500 units/g agarose (One ml gel will yield 65-120 units)