Merck
Covalent α-synuclein dimers: chemico-physical and aggregation properties.
PloS one (2012-12-29)
Micaela Pivato, Giorgia De Franceschi, Laura Tosatto, Erica Frare, Dhruv Kumar, Daniel Aioanei, Marco Brucale, Isabella Tessari, Marco Bisaglia, Bruno Samori, Patrizia Polverino de Laureto, Luigi Bubacco
ZUSAMMENFASSUNG
The aggregation of α-synuclein into amyloid fibrils constitutes a key step in the onset of Parkinson's disease. Amyloid fibrils of α-synuclein are the major component of Lewy bodies, histological hallmarks of the disease. Little is known about the mechanism of aggregation of α-synuclein. During this process, α-synuclein forms transient intermediates that are considered to be toxic species. The dimerization of α-synuclein could represent a rate-limiting step in the aggregation of the protein. Here, we analyzed four covalent dimers of α-synuclein, obtained by covalent link of the N-terms, C-terms, tandem cloning of two sequences and tandem juxtaposition in one protein of the 1-104 and 29-140 sequences. Their biophysical properties in solution were determined by CD, FT-IR and NMR spectroscopies. SDS-induced folding was also studied. The fibrils formation was analyzed by ThT and polarization fluorescence assays. Their morphology was investigated by TEM and AFM-based quantitative morphometric analysis. All dimers were found to be devoid of ordered secondary structure under physiological conditions and undergo α-helical transition upon interaction with SDS. All protein species are able to form amyloid-like fibrils. The reciprocal orientation of the α-synuclein monomers in the dimeric constructs affects the kinetics of the aggregation process and a scale of relative amyloidogenic propensity was determined. Structural investigations by FT IR spectroscopy, and proteolytic mapping of the fibril core did not evidence remarkable difference among the species, whereas morphological analyses showed that fibrils formed by dimers display a lower and diversified level of organization in comparison with α-synuclein fibrils. This study demonstrates that although α-synuclein dimerization does not imply the acquisition of a preferred conformation by the participating monomers, it can strongly affect the aggregation properties of the molecules. The results presented highlight a substantial role of the relative orientation of the individual monomer in the definition of the fibril higher structural levels.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Proteinase K aus Tritirachium album, lyophilized powder, ≥30 units/mg protein
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Proteinase K aus Tritirachium album, lyophilized powder, Molecular Biology, BioUltra, ≥30 units/mg protein
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Proteinase K aus Tritirachium album, buffered aqueous glycerol solution, Molecular Biology, ≥800 units/mL
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Proteinase K aus Tritirachium album, ≥3.0 unit/mg solid, lyophilized powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Proteinase K aus Tritirachium album, ≥500 units/mL, buffered aqueous glycerol solution
Preise und Verfügbarkeit sind derzeit nicht verfügbar.