Merck
  • Poly (ADP-Ribose) polymerase inhibitor MK-4827 together with radiation as a novel therapy for metastatic neuroblastoma.
Poly (ADP-Ribose) polymerase inhibitor MK-4827 together with radiation as a novel therapy for metastatic neuroblastoma.
Anticancer research (2013-03-14)
Sabine Mueller, Samhita Bhargava, Annette M Molinaro, Xiaodong Yang, Ilan Kolkowitz, Aleksandra Olow, Noor Wehmeijer, Sharon Orbach, Justin Chen, Katherine K Matthay, Daphne A Haas-Kogan
ZUSAMMENFASSUNG
To assess poly (ADP-ribose) polymerase (PARP) inhibitor MK-4827 together with radiation for the treatment of neuroblastoma. Clonogenic survival assays were used to assess MK-4827, radiation and combination thereof in four neuroblastoma cell lines. In vivo efficacy was tested in a murine xenograft model of metastatic neuroblastoma. In vivo targeted inhibition and biological effects included measurement of cleaved caspase-3, γ-H2AX, and Ki 67 by immunohistochemistry (IHC) and poly-ADP-ribose by Enzyme-Linked Immunosorbent Assay. Treatment of neuroblastoma cell lines reduced clonogenicity and resulted in additive effects with radiation. In vivo treatment with MK-4827 and radiation prolonged survival (p<0.01) compared to single modalities. In vivo superiority of MK-4827 plus radiation was further documented by significant elevations of cleaved caspase-3 and γ-H2AX in tumors from the combination group compared to single modality cohorts. Combination of MK-4827 and radiation might provide effective therapy for children with high-risk neuroblastoma.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
PARP-1 human, recombinant, expressed in E. coli
Preise und Verfügbarkeit sind derzeit nicht verfügbar.