Merck
  • Ethyl pyruvate significantly inhibits tumour necrosis factor-α, interleukin-1β and high mobility group box 1 releasing and attenuates sodium taurocholate-induced severe acute pancreatitis associated with acute lung injury.
Ethyl pyruvate significantly inhibits tumour necrosis factor-α, interleukin-1β and high mobility group box 1 releasing and attenuates sodium taurocholate-induced severe acute pancreatitis associated with acute lung injury.
Clinical and experimental immunology (2013-04-23)
Z-G Luan, J Zhang, X-H Yin, X-C Ma, R-X Guo
ZUSAMMENFASSUNG
In this study, we examined the effect of ethyl pyruvate (EP) on pulmonary inflammation in rats with severe pancreatitis-associated acute lung injury (ALI). Severe acute pancreatitis (SAP) was induced in rats by the retrograde injection of 5% sodium taurocholate into the pancreatic duct. Rats were randomly divided into the following experimental groups: control group, SAP group and EP-treated group. The tissue specimens were harvested for morphological studies, Streptavidin-peroxidase immunohistochemistry examination. Pancreatic or lung tissue oedema was evaluated by tissue water content. Serum amylase and lung tissue malondialdehyde (MDA) and myeloperoxidase (MPO) were measured. Meanwhile, the nuclear factor-κB (NF-κB) activation, tumour necrosis factor-α (TNF-α), interleukin-1β (IL-1β) levels and HMGB1 protein expression levels in the lung were studied. In the present study, we demonstrated that treatment with EP after SAP was associated with a reduction in the severity of SAP and lung injury. Treatment with EP significantly decreased the expression of TNF-α, IL-1β, HMGB1 and ameliorated MDA concentration, MPO activity in the lung in SAP rats. Compared to SAP group, administration of EP prevented pancreatitis-induced increases in nuclear translocation of NF-κB in the lung. Similarly, treatment with EP significantly decreased the accumulation of neutrophils and markedly reduced the enhanced lung permeability. In conclusion, these results demonstrate that EP might play a therapeutic role in pulmonary inflammation in this SAP model.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Peroxidase aus Meerrettich, Type VI, essentially salt-free, lyophilized powder, ≥250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Peroxidase aus Meerrettich, Type VI-A, essentially salt-free, lyophilized powder, 950-2000 units/mg solid (using ABTS), ≥250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Peroxidase aus Meerrettich, Type II, essentially salt-free, lyophilized powder, 150-250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Peroxidase aus Meerrettich, lyophilized, powder, ~150 U/mg
Sigma-Aldrich
Peroxidase aus Meerrettich, Type I, essentially salt-free, lyophilized powder, ≥50 units/mg solid (using pyrogallol)
Sigma-Aldrich
Myeloperoxidase aus Humanleukocyten, lyophilized powder, ≥50 units/mg protein
Sigma-Aldrich
Ethylpyruvat, 98%
Sigma-Aldrich
Peroxidase aus Meerrettich, Highly stabilized, essentially salt-free, lyophilized powder, 200-300 units/mg solid (using pyrogallol)
Sigma-Aldrich
Lactoperoxidase aus Kuhmilch, lyophilized powder (essentially salt-free), ≥200 units/mg protein
Natriumtaurocholat, BRP, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Peroxidase aus Meerrettich, Type X, ammonium sulfate suspension
Sigma-Aldrich
Peroxidase aus Meerrettich, Type XII, essentially salt-free, lyophilized powder, ≥250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Ethylpyruvat, natural, FG
Sigma-Aldrich
Lactoperoxidase aus Kuhmilch, lyophilized, powder, ≥150 U/mg