Merck
Efficient production of ethanol from crude glycerol by a Klebsiella pneumoniae mutant strain.
Bioresource technology (2010-12-28)
Baek-Rock Oh, Jeong-Woo Seo, Sun-Yeon Heo, Won-Kyung Hong, Lian Hua Luo, Min-ho Joe, Don-Hee Park, Chul Ho Kim
ZUSAMMENFASSUNG
A mutant strain of Klebsiella pneumoniae, termed GEM167, was obtained by γ irradiation, in which glycerol metabolism was dramatically affected on exposure to γ rays. Levels of metabolites of the glycerol reductive pathway, 1,3-propanediol (1,3-PD) and 3-hydroxypropionic acid (3-HP), were decreased in the GEM167 strain compared to a control strain, whereas the levels of metabolites derived from the oxidative pathway, 2,3-butanediol (2,3-BD), ethanol, lactate, and succinate, were increased. Notably, ethanol production from glycerol was greatly enhanced upon fermentation by the mutant strain, to a maximum production level of 21.5 g/l, with a productivity of 0.93 g/l/h. Ethanol production level was further improved to 25.0 g/l upon overexpression of Zymomonas mobilis pdc and adhII genes encoding pyruvate decarboxylase (Pdc) and aldehyde dehydrogenase (Adh), respectively in the mutant strain GEM167.