Merck
  • Activation of SIRT1 by resveratrol induces KLF2 expression conferring an endothelial vasoprotective phenotype.
Activation of SIRT1 by resveratrol induces KLF2 expression conferring an endothelial vasoprotective phenotype.
Cardiovascular research (2009-10-10)
Jorge Gracia-Sancho, Guadalupe Villarreal, Yuzhi Zhang, Guillermo García-Cardeña
ZUSAMMENFASSUNG
Resveratrol activates Sirtuin 1 (SIRT1), a nicotinamide adenine dinucleotide-dependent deacetylase which modulates metabolic homeostasis and improves several pathophysiological features present in diseases of ageing. In particular, it has been shown that SIRT1 activation improves endothelial dysfunction and suppresses vascular inflammation, two central pathophysiological processes involved in the initiation and progression of cardiovascular disease. The downstream targets of SIRT1 activation in this context, however, remain poorly defined. Therefore, in this study, we aimed to characterize mechanistically how SIRT1 activation regulates the endothelial vasoprotective phenotype. We demonstrate that SIRT1 activation by resveratrol increases the expression of the transcription factor Krüppel-like factor 2 (KLF2) in human vascular endothelial cells, resulting in the orchestrated regulation of transcriptional programs critical for conferring an endothelial vasoprotective phenotype. Moreover, we show that KLF2 upregulation by resveratrol occurs via a mitogen-activated protein kinase 5/myocyte enhancing factor 2-dependent signalling pathway. Collectively, these observations provide a new mechanistic framework to understand the vascular protective effects mediated by SIRT1 activators and define KLF2 as a critical mediator of these effects.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
AMPK-Inhibitor, Verbindung C, AMPK Inhibitor, Compound C, CAS 866405-64-3, is a cell-permeable compound that inhibits KDR/VEGFR2, ALK2/BMPR-I, and AMPK kinase activities (IC50 = 25.1, 148, and 234.6 nM, respectively).
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Sirtinol, ≥95% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.