Merck
  • Regulation of the EphA2 kinase by the low molecular weight tyrosine phosphatase induces transformation.
Regulation of the EphA2 kinase by the low molecular weight tyrosine phosphatase induces transformation.
The Journal of biological chemistry (2002-08-09)
Keith D Kikawa, Derika R Vidale, Robert L Van Etten, Michael S Kinch
ZUSAMMENFASSUNG
Intracellular signaling by protein tyrosine phosphorylation is generally understood to govern many aspects of cellular behavior. The biological consequences of this signaling pathway are important because the levels of protein tyrosine phosphorylation are frequently elevated in cancer cells. In the classic paradigm, tyrosine kinases promote tumor cell growth, survival, and invasiveness, whereas tyrosine phosphatases negatively regulate these same behaviors. Here, we identify one particular tyrosine phosphatase, low molecular weight tyrosine phosphatase (LMW-PTP), which is frequently overexpressed in transformed cells. We also show that overexpression of LMW-PTP is sufficient to confer transformation upon non-transformed epithelial cells. Notably, we show that the EphA2 receptor tyrosine kinase is a prominent substrate for LMW-PTP and that the oncogenic activities of LMW-PTP result from altered EphA2 expression and function. These results suggest a role for LMW-PTP in transformation progression and link its oncogenic potential to EphA2.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phosphatase, sauer aus Kartoffeln, lyophilized powder, ≥0.5 unit/mg solid
Sigma-Aldrich
Anti-Phosphotyrosin-Antikörper, Klon 4G10®, clone 4G10®, Upstate®, from mouse
Sigma-Aldrich
Phosphatase, sauer aus Weizenkeimen, ≥0.4 unit/mg solid
Sigma-Aldrich
Phosphatase, sauer aus Kartoffeln, lyophilized powder, ≥3.0 units/mg solid
Sigma-Aldrich
Phosphatase, sauer aus Süsskartoffeln, ammonium sulfate suspension, ≥10.0 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Anti-Eck/EphA2-Antikörper, Klon D7, clone D7, Upstate®, from mouse