Merck
  • Improving MCE with electrochemical detection using a bubble cell and sample stacking techniques.
Improving MCE with electrochemical detection using a bubble cell and sample stacking techniques.
Electrophoresis (2009-10-06)
Qian Guan, Charles S Henry
ZUSAMMENFASSUNG
Two efforts to improve the sensitivity and limits of detection for MCE with electrochemical detection are presented here. One is the implementation of a capillary expansion (bubble cell) at the detection zone to increase the exposed working electrode surface area. Bubble cell widths were varied from 1x to 10x the separation channel width (50 mum) to investigate the effects of electrode surface area on detection sensitivity, LOD, and separation efficiency. Improved detection sensitivity and decreased detection limits were obtained with increased bubble cell width, and LODs of dopamine and catechol detected in a 5x bubble cell were 25 and 50 nM, respectively. Meanwhile, fluorescent imaging results demonstrated approximately 8 and approximately 12% loss in separation efficiency in 4x and 5x bubble cell, respectively. Another effort at reducing the LOD involves using field amplified sample injection for gated injection and field amplified sample stacking for hydrodynamic injection. Stacking effects are shown for both methods using amperometric detection and pulsed amperometric detection. The LODs of dopamine in a 4x bubble cell were 8 and 20 nM using field amplified sample injection and field amplified sample stacking, respectively. However, improved LODs were not obtained for anionic analytes using either stacking technique.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Cystein, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
L-Glutathion reduziert, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathion reduziert, ≥98.0%
Sigma-Aldrich
L-Cystein, 97%
Sigma-Aldrich
L-Cystein, BioUltra, ≥98.5% (RT)
SAFC
L-Cystein
Sigma-Aldrich
L-Glutathion reduziert, BioXtra, ≥98.0%
Sigma-Aldrich
L-Cystein, ≥97%, FG
Supelco
L-Cystein, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Cystein, Wacker Chemie AG, ≥98.0%