Merck
  • High-performance liquid chromatography and inductively coupled plasma mass spectrometry (HPLC-ICP-MS) for the analysis of xenobiotic metabolites in rat urine: application to the metabolites of 4-bromoaniline.
High-performance liquid chromatography and inductively coupled plasma mass spectrometry (HPLC-ICP-MS) for the analysis of xenobiotic metabolites in rat urine: application to the metabolites of 4-bromoaniline.
The Analyst (2000-05-23)
J K Nicholson, J C Lindon, G Scarfe, I D Wilson, F Abou-Shakra, J Castro-Perez, A Eaton, S Preece
ZUSAMMENFASSUNG
The use of HPLC-ICP-MS for the profiling and quantification of the metabolites of 4-bromoaniline following reversed-phase gradient chromatography is demonstrated. In the 0-8 h post dose sample, which contained the highest concentrations of compound-related material, it was possible to detect at least 16 metabolites of the compound. The methodology described offers the possibility of obtaining metabolite profiles and quantification for drugs and other xenobiotics in biological fluids and excreta without the requirement for radiolabelled tracers.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Bromanilin, 97%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
4-Bromanilin, ≥99.0% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.