Merck
Soa genotype selectively affects mouse gustatory neural responses to sucrose octaacetate.
Physiological genomics (2001-05-01)
M Inoue, X Li, S A McCaughey, G K Beauchamp, A A Bachmanov
ZUSAMMENFASSUNG
In mice, behavioral acceptance of the bitter compound sucrose octaacetate (SOA) depends on allelic variation of a single gene, Soa. The SW.B6-Soa(b)congenic mouse strain has the genetic background of an "SOA taster" SWR/J strain and an Soa-containing donor chromosome fragment from an "SOA nontaster" C57BL/6J strain. Using microsatellite markers polymorphic between the two parental strains, we determined that the donor fragment spans 5-10 cM of distal chromosome 6. The SWR/J mice avoided SOA in two-bottle tests with water and had strong responses to SOA in two gustatory nerves, the chorda tympani (CT) and glossopharyngeal (GL). In contrast, the SW.B6-Soa(b) mice were indifferent to SOA in two-bottle tests and had very weak responses to SOA in both of these nerves. The SWR/J and SW.B6-Soa(b) mice did not differ in responses of either nerve to sucrose, NaCl, HCl, or the bitter-tasting stimuli quinine, denatonium, strychnine, 6-n-propylthiouracil, phenylthiocarbamide, and MgSO(4). Thus the effect of the Soa genotype on SOA avoidance is mediated by peripheral taste responsiveness to SOA, involving taste receptor cells innervated by both the CT and GL nerves.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Sucroseoctaacetat, ≥97%, FG
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
D-(+)-Saccharoseoctaacetat, 98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.