Merck
  • Flame atomic absorption spectrometric determination of trace amounts of manganese in alloys and biological samples after preconcentration with the ion pair of 2-(5-bromo-2-pyridylazo)-5-diethylaminophenol and ammonium tetraphenylborate on microcrystalline naphthalene or by column method.
Flame atomic absorption spectrometric determination of trace amounts of manganese in alloys and biological samples after preconcentration with the ion pair of 2-(5-bromo-2-pyridylazo)-5-diethylaminophenol and ammonium tetraphenylborate on microcrystalline naphthalene or by column method.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2001-11-16)
M A Taher
ZUSAMMENFASSUNG
Manganese is quantitatively retained on 2-(5-bromo-2-pyridylazo)-5-diethylaminophenol (5-Br-PADAP)-ammonium tetraphenylborate with microcrystalline naphthalene or by a column method in the pH range 7.5-10.5 from large volumes of aqueous solutions of various samples. After filtration, each solid mass consisting of the manganese complex and naphthalene was dissolved with 5 ml of dimethylformamide and the metal was determined by flame atomic absorption spectrometry. Manganese complex can alternatively be quantitatively adsorbed on ammonium tetraphenylborate-naphthalene adsorbent packed in a column and determined similarly. About 0.1 microgram of manganese can be concentrated in a column from 500 ml of aqueous sample, where its concentration is as low as 0.2 ppb. Eight replicate determinations of 1.0 ppm of manganese gave a mean absorbance of 0.224 with a relative standard deviation of 1.8%. The sensitivity for 1% absorption was 19 ppb. The interference of a large number of anions and cations has been studied and the optimized conditions developed were utilized for the trace determination of manganese in various standard samples.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-(5-Brom-2-pyridylazo)-5-(diethylamino)phenol, 97%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.