Merck
  • Allergic contact dermatitis from cocamidopropyl betaine, cocamidoamine, 3-(dimethylamino)propylamine, and oleamidopropyl dimethylamine: co-reactions or cross-reactions?
Allergic contact dermatitis from cocamidopropyl betaine, cocamidoamine, 3-(dimethylamino)propylamine, and oleamidopropyl dimethylamine: co-reactions or cross-reactions?
Dermatitis : contact, atopic, occupational, drug (2005-02-24)
Linda Moreau, Denis Sasseville
ZUSAMMENFASSUNG
We present the case of a patient with facial dermatitis caused by sensitization to cocamidopropyl betaine. The patient also had positive patch-test reactions to cocamidoamine, 3-(dimethylamino)propylamine, and oleamidopropyl dimethylamine. The presence of 3-(dimethylamino)propylamine as an impurity in all of these substances can be hypothesized to explain these simultaneous reactions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-(Dimethylamino)-1-Propylamin, 99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
3-(Dimethylamino)-1-Propylamin, purum, ≥98.0% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.