Merck
A rapid and sensitive GC-MS method for determination of 1,3-dichloro-2-propanol in water.
Analytical and bioanalytical chemistry (2005-04-28)
Rainer Schuhmacher, Johanna Nurmi-Legat, Andreas Oberhauser, Markus Kainz, Rudolf Krska
ZUSAMMENFASSUNG
A rapid analytical method for sensitive determination of 1,3-dichloro-2-propanol (1,3-DCP) in river water has been developed. 1,3-DCP is extracted from water with ethyl acetate. After filtration through sodium sulfate the ethyl acetate phase is analyzed by gas chromatography-mass spectrometry. The method uses 1,3-DCP-d5 as internal standard. Different extraction solvents, concentrations of ammonium sulfate in the water samples, and the effect of humic acid were tested and their influence on the recovery of DCP has been evaluated. The method quantification limit was 0.1 microg L(-1). For spiked water samples (0-5.2 microg L(-1), n=21) a repeatability coefficient of variation of 5.4% was obtained. The average recovery rate of 1,3-DCP was 105+/-3% (n=21). Stability tests, which were carried out with Danube river water, led to an estimated 1,3-DCP degradation rate of 0.008+/-0.0008 day(-1) at 6 degrees C.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,3-Dichlor-2-propanol, 97%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.