Merck
Signaling molecularly imprinted polymers: molecular recognition-based sensing materials.
Chemical record (New York, N.Y.) (2005-10-08)
Toshifumi Takeuchi, Takashi Mukawa, Hideyuki Shinmori
ZUSAMMENFASSUNG
Molecular imprinting is a template polymerization technique that can easily provide synthetic polymers capable of molecular recognition for given target molecules. In addition to their highly specific recognition ability, we are attempting to introduce signaling functions to molecularly imprinted polymers, enabling them to respond according to specific binding events. Some of our work regarding such signaling molecularly imprinted polymers is presented here, including molecularly imprinted polymers that induce spectral shifts of target compounds because of binding. Such compounds include hydrogen-bonding-based fluorescent imprinted polymers and metalloporphyrin-based signaling molecularly imprinted polymers.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cinchonidin, 96%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.