Merck
  • Lack of effectiveness of several chelators in removing internally deposited strontium from mice following repeated parenteral strontium administration.
Lack of effectiveness of several chelators in removing internally deposited strontium from mice following repeated parenteral strontium administration.
Veterinary and human toxicology (1992-02-01)
J M Llobet, M T Colomina, J L Domingo, J Corbella
ZUSAMMENFASSUNG
Diethylenetriaminepentaacetic acid (DTPA), ethylenglycolbis-(beta-amino-ethylether)-N,N-tetraacetic acid (EGTA), tartaric acid, KRYPTOFIX 222, and KRYPTOFIX 5 were evaluated for their efficacy in mobilization of strontium from the body of mice which had received 20 sc injections of strontium nitrate (95 mg/kg/injection) for 4 w. Twenty-four hours after the last strontium injection, ip administration of 1 of the various chelators or 0.9% saline was initiated and continued daily for 5 d. Mice were housed in metabolic cages, and urine and feces were collected daily for 5 d. After this period, the animals were killed and tissues removed. Tartaric acid, KRYPTOFIX 222, and KRYPTOFIX 5 had no effect on urinary or fecal strontium elimination, whereas DTPA and EGTA significantly decreased the fecal strontium excretion. The concentration of strontium in bone was only lowered in tartaric-treated mice. This study indicates the use of the above chelators is not an effective treatment to enhance the removal of strontium following repeated parenteral strontium administration.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosan, 98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.