Merck
Congo red interaction with alpha-proteins.
Acta biochimica Polonica (2006-03-28)
Jolanta Sereikaite, Vladas-Algirdas Bumelis
ZUSAMMENFASSUNG
The ability of Congo red to form complexes with alpha-proteins, human growth hormone and human interferon-alpha2b, was found by absorption difference spectroscopy. A human growth hormone-Congo red complex was isolated by gel-permeation chromatography, and its visible absorption spectrum was registered in comparison to free dye. The ability of Congo red to induce dimerization of human growth hormone was demonstrated using chemical cross-linking agents 1,3,5-triacryloyl-hexahydro-s-triazine and ethylene glycol bis(succinimidylsuccinate).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,3,5-Triacryloylhexahydro-1,3,5-triazin, 98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethylenglykol-disuccinat-di-(N-succinimidyl)-ester, powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.