Merck
  • Novel carbohydrate-binding activity of bovine liver beta-glucuronidase toward lactose/N-acetyllactosamine sequences.
Novel carbohydrate-binding activity of bovine liver beta-glucuronidase toward lactose/N-acetyllactosamine sequences.
Glycobiology (2006-06-16)
Hiroko Matsushita-Oikawa, Mayumi Komatsu, Naoko Iida-Tanaka, Hiromi Sakagami, Tetsuko Kanamori, Isamu Matsumoto, Nobuko Seno, Haruko Ogawa
ZUSAMMENFASSUNG
Beta-glucuronidase is a lysosomal enzyme that plays an essential role in normal turnover of glycosaminoglycans and remodeling of the extracellular matrix components in both physiological and inflammatory states. The regulation mechanisms of enzyme activity and protein targeting of beta-glucuronidase have implications for the development of a variety of therapeutics. In this study, the effectiveness of various carbohydrate-immobilized adsorbents for the isolation of bovine liver beta-glucuronidase (BLG) from other glycosidases was tested. Beta-glucuronidase and contaminating glycosidases in commercial BLG preparations bound to and were coeluted from adsorbents immobilized with the substrate or an inhibitor of beta-glucuronidase, whereas beta-glucuronidase was found to bind exclusively with lactamyl-Sepharose among the adsorbents tested and to be effectively separated from other enzymes. Binding and elution studies demonstrated that the interaction of beta-glucuronidase with lactamyl-Sepharose is pH dependent and carbohydrate specific. BLG was purified to homogeneity by lactamyl affinity chromatography and subsequent anion-exchange high-performance liquid chromatography (HPLC). Lactose was found to activate beta-glucuronidase noncompetitively, indicating that the lactose-binding site is different from the substrate-binding site. Binding studies with biotinyl glycoproteins, lipids, and synthetic sugar probes revealed that beta-glucuronidase binds to N-acetyllactosamine/lactose-containing glycoconjugates at neutral pH. The results indicated the presence of N-acetyllactosamine/lactose-binding activity in BLG and provided an effective purification method utilizing the novel carbohydrate binding activity. The biological significance of the carbohydrate-specific interaction of beta-glucuronidase, which is different from the substrate recognition, is discussed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
β-Glucuronidase aus Helix pomatia, Type HP-2, aqueous solution, ≥100,000 units/mL
Sigma-Aldrich
ββ-Glucuronidase aus E. coli, Type IX-A, lyophilized powder, 1,000,000-5,000,000 units/g protein (30 min assay)
Sigma-Aldrich
β-Glucuronidase aus Napfschnecke (Patella vulgata), Type L-II, lyophilized powder, 1,000,000-3,000,000 units/g solid
Sigma-Aldrich
ββ-Glucuronidase aus E. coli, Type VII-A, lyophilized powder, 5,000,000-20,000,000 units/g protein, pH 6.8 (30 min assay)
Sigma-Aldrich
β-Glucuronidase aus Helix pomatia, Type H-2, aqueous solution, ≥85,000 units/mL
Sigma-Aldrich
β-Glucuronidase aus Helix pomatia, Type H-1, partially purified powder, ≥300,000 units/g solid
Sigma-Aldrich
β-Glucuronidase aus Rinderleber, Type B-1, ≥1,000,000 units/g solid
Sigma-Aldrich
β-Glucuronidase aus Helix pomatia, Type H-5, lyophilized powder, ≥400,000 units/g solid
Sigma-Aldrich
β-Glucuronidase aus Helix pomatia, Type H-3, aqueous solution, ≥90,000 units/mL
Sigma-Aldrich
β-Glucuronidase aus Helix pomatia, Type HP-2S, aqueous solution, ≥90,000 units/mL
Sigma-Aldrich
ββ-Glucuronidase aus E. coli, ≥10,000,000 units/g protein (30 min assay), recombinant, expressed in E. coli overproducing strain, lyophilized powder
Sigma-Aldrich
Diethylaminoethyl–Sephacel®, aqueous ethanol suspension, 40-160 μm (wet), exclusion limit ~1,000,000 Da
Sigma-Aldrich
ββ-Glucuronidase aus E. coli, ≥20,000 units/mg protein, recombinant, expressed in E. coli overproducing strain, lyophilized powder
Sigma-Aldrich
ββ-Glucuronidase aus E. coli, aqueous glycerol solution, ≥5,000,000 units/g protein, pH 6.8 (biuret)
Sigma-Aldrich
β-Glucuronidase aus Rinderleber, Type B-3, ≥2,000,000 units/g solid
Sigma-Aldrich
β-Glucuronidase aus Napfschnecke (Patella vulgata), aqueous solution, ≥85,000 units/mL
Sigma-Aldrich
β-Glucuronidase aus Helix pomatia, Type H-3AF, aqueous solution, ≥60,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix aspersa (garden snail), Type HA-4