Merck
  • Ab initio calculations on SCl2 and low-lying cationic states of SCl2+: Franck-Condon simulation of the UV photoelectron spectrum of SCl2.
Ab initio calculations on SCl2 and low-lying cationic states of SCl2+: Franck-Condon simulation of the UV photoelectron spectrum of SCl2.
The Journal of chemical physics (2006-09-27)
Daniel K W Mok, Foo-Tim Chau, Edmond P F Lee, John M Dyke
ZUSAMMENFASSUNG
Geometry optimization calculations were carried out on the (approximate)X (1)A(1) state of SCl(2) and the (approximate)X(2)B(1), (approximate)A(2)B(2), (approximate)B(2)A(1), (approximate)C(2)A(1), (approximate)D(2)A(2), and (approximate)E (2)B(2) states of SCl(2) (+) at the restricted-spin coupled-cluster single-double plus perturbative triple excitation [RCCSD(T)] level with basis sets of up to the augmented correlation-consistent polarized quintuple-zeta [aug-cc-pV(5+d)Z] quality. Effects of core electron correlation, basis set extension to the complete basis set limit, and relativistic contributions on computed minimum-energy geometrical parameters and/or relative electronic energies were also investigated. RCCSD(T) potential energy functions (PEFs) were calculated for the (approximate)X (1)A(1) state of SCl(2) and the low-lying states of SCl(2)(+) listed above employing the aug-cc-pV(5+d)Z basis set. Anharmonic vibrational wave functions of these neutral and cationic states of SCl(2), and Franck-Condon (FC) factors of the lowest four one-electron allowed neutral photoionizations were computed employing the RCCSD(T)aug-cc-pV(5+d)Z PEFs. Calculated FC factors with allowance for the Duschinsky rotation and anharmonicity were used to simulate the first four photoelectron (PE) bands of SCl(2). The agreement between simulated and observed He I PE spectra reported by Colton et al. [J. Electron Spectrosc. Relat. Phenom. 3, 345 (1974)] and Solouki et al. [Chem. Phys. Lett. 26, 20 (1974)] is excellent. However, our FC spectral simulations indicate that the first observed vibrational component in the first PE band of SCl(2) is a "hot" band arising from the SCl(2)(+)(approximate)X(2)B(1)(0,0,0)<--SCl(2)(approximate)X (1)A(1)(1,0,0) ionization. Consequently, the experimental adiabatic ionization energy of SCl(2) is revised to 9.55+/-0.01 eV, in excellent agreement with results obtained from state-of-the-art ab initio calculations in this work.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Schwefelmonochlorid, 98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.