Merck
  • Flavor authentication studies of alpha-ionone, beta-ionone, and alpha-ionol from various sources.
Flavor authentication studies of alpha-ionone, beta-ionone, and alpha-ionol from various sources.
Journal of agricultural and food chemistry (2007-07-17)
María del Mar Caja, Christina Preston, Michael Kempf, Peter Schreier
ZUSAMMENFASSUNG
In addition to the already available information on the authenticity of alpha- (1) and beta-ionone (2) from plant tissues, there is an interest in the stable isotope data of 1 and 2 available by synthesis from citral and acetone, as European Union regulations, in contrast to the United States and other countries, do not allow a product to be declared as 'natural' that has been chemically synthesized (e.g., by using a natural catalyst) from natural educts. Analyses performed by on-line capillary gas chromatography-isotope ratio mass spectrometry in the combustion and pyrolysis modes (HRGC-C/P-IRMS) as well as by elemental analyzers (EA-C/P-IRMS) measuring delta(13)C(V)-PDB and delta(2)H(V)-SMOW values provide for the first time isotope data of such 'natural' 1 and 2 as well as of synthetic and 'ex plant' alpha-ionol (3). The isotope data recorded for synthesized 1 and 2 reflected the influence of the origin of the used citral, whereas that of acetone was less remarkable. For instance, 'natural' 1 ex citral from lemongrass showed, as expected for a C4 plant, an enriched delta(13)C(V)-PDB value of -18.5 per thousand. In addition, the use of synthetic citral resulted in an enriched delta(2)H(V)-SMOW value of -43 per thousand, whereas with citral ex Litsea cubeba and ex lemongrass values of -242 and -232 per thousand, respectively, were recorded. IRMS analyses of 'natural' 2 revealed delta(13)C(V)-PDB and delta(2)H(V)-SMOW values that were nearly identical to that recorded for 'natural' 1. As to both 1 and 2, variations of synthesis conditions led to distinct changes in the delta(13)C(V)-PDB but not the delta(2)H(V)-SMOW values. Synthetic 3 showed delta(13)C(V)-PDB and delta(2)H(V)-SMOW values of -24.5 and -184 per thousand, respectively. These data differed from those found in raspberry fruit under study (n = 8), that is, ranging from -33.6 to -36.6 per thousand for delta(13)C(V)-PDB and from -200 to -225 per thousand for delta(2)H(V)-SMOW. The values determined additionally for 1 and 2 in raspberry fruit samples ranged from -30.3 to -35.1 per thousand and from -176 to -221 per thousand for delta(13)C(V)-PDB and delta(2)H(V)-SMOW, respectively, and thus corresponded to the already known literature information.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
α-Ionon, ≥90%, stabilized
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
α-Ionon, technical grade, 90%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.