Merck
Antidepressant-like effect of agmatine is not mediated by serotonin.
Behavioural brain research (2008-01-08)
Maarja Krass, Gregers Wegener, Eero Vasar, Vallo Volke
ZUSAMMENFASSUNG
The aim of this study was to characterize the behavioral effects of systemically administered agmatine in animal models predictive of antidepressant- and anxiolytic-like activity and clarify whether the effects of agmatine depend on the intact serotonergic system. Only the highest dose of agmatine tested (50 mg/kg) decreased immobility of mice in the forced swimming test. The magnitude of the effect was slightly smaller than that of the tricyclic antidepressant imipramine (15 mg/kg). Agmatine did not change the locomotion of mice in the open field. Pretreatment with the tryptophane hydroxylase inhibitor PCPA for 3 days resulted in more than 70% drop in the tissue levels of 5-HT and 5-HIIA but did not counteract the antidepressant-like effect of agmatine. The administration of agmatine did not modify behavior of animals in the light-dark compartment test of anxiety. We conclude that the antidepressant-like effect of agmatine seems not to be mediated by the serotonergic system. We failed to confirm the reported anxiolytic-like activity of agmatine.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Chlor-DL-Phenylalanin-Methylester -hydrochlorid
Preise und Verfügbarkeit sind derzeit nicht verfügbar.