Merck
  • Electronic structures of azulene-fused porphyrins as seen by magnetic circular dichroism and TD-DFT calculations.
Electronic structures of azulene-fused porphyrins as seen by magnetic circular dichroism and TD-DFT calculations.
Journal of inorganic biochemistry (2008-01-09)
Katsunori Nakai, Kei Kurotobi, Atsuhiro Osuka, Masanobu Uchiyama, Nagao Kobayashi
ZUSAMMENFASSUNG
A combination of magnetic circular dichroism (MCD), electronic absorption spectroscopy and time-dependent density functional theory (TD-DFT) calculations has been used to investigate the electronic structure of azulene-fused pi-expanded porphyrins based primarily on the spectral properties of absorption bands in the near infrared region. From MCD experiments, it was suggested that in the case of a mono-azulene-fused porphyrin DeltaHOMO approximately equal DeltaLUMO (where DeltaHOMO is the magnitude of the energy gap between the HOMO and HOMO-1 and DeltaLUMO is the magnitude of the energy gap between the LUMO and LUMO+1), while in the case of an oppositely-di-azulene-fused porphyrin, DeltaHOMO<DeltaLUMO. Since Faraday A terms are observed for both the Soret and Q bands in the MCD spectrum of tetra-azulene-fused porphyrin the corresponding excited states are clearly accidentally degenerate despite the C(2) molecular symmetry. Transition dipole moment analysis clearly demonstrates that the electronic absorption spectrum of tetra-azulene-fused porphyrin has out-of-plane electronic transitions slightly to the blue of the main Q and Soret bands. Comparison with distorted porphyrins and phthalocyanines strongly suggests that these out-of-plane transitions appear as intense Gaussian-shaped Faraday B terms in the MCD spectra.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Azulen, 99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.