Merck
  • Determination of antibacterial quaternary ammonium compound in lozenges and human serum by resonance light scattering technique.
Determination of antibacterial quaternary ammonium compound in lozenges and human serum by resonance light scattering technique.
Journal of pharmaceutical and biomedical analysis (2008-07-01)
Zhanguang Chen, Yurui Peng, Junhui Chen, Li Zhu
ZUSAMMENFASSUNG
A method for the specific determination of an antibacterial quaternary ammonium compound Dequalinium chloride (DQC) was described in this paper. At pH 0.5, the resonance light scattering (RLS) intensity of sodium dodecyl benzene sulfonate (SDBS) remarkably was enhanced by adding DQC. A RLS peak at 392.0 nm was found, and the enhanced intensity of RLS at this wavelength was proportional to the concentration of DQC in the range of 0.096-2.88 microg/mL. The detection limit was 2.98 ng/mL and the correlation coefficient was r=0.9988 (n=9). The method was applied to the analysis of DQC in lozenges and human serum. The results indicated that the method was sensitive, simple, practical and useful in the clinical assay.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Dequaliniumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.