Merck
  • Structural and spectroscopic elucidation of tetrapetide glycyl-(L)-prolyl-glycyl-glycine and its hydrogensquarate.
Structural and spectroscopic elucidation of tetrapetide glycyl-(L)-prolyl-glycyl-glycine and its hydrogensquarate.
Protein and peptide letters (2008-09-11)
Tsonko M Kolev
ZUSAMMENFASSUNG
The IR-spectroscopic and structural elucidation of tetrapeptide glycyl-(L)-prolyl-glycyl-glycine and its hydrogensquarate was performed by employing linear-polarized IR-spectroscopy of oriented colloid suspensions in nematic host as well as mass spectrometry. Quantum-chemical ab initio calculations were carried out in order to evaluate both the electronic structure and optical properties of the compound studied.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3,4-Dihydroxy-3-cyclobuten-1,2-dion, ≥99.0% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.