Merck
  • Low levels of gibberellic acid control the biosynthesis of ajmalicine in Catharanthus roseus cell suspension cultures.
Low levels of gibberellic acid control the biosynthesis of ajmalicine in Catharanthus roseus cell suspension cultures.
Planta medica (2008-12-20)
Aniça Amini, Gaëlle Glévarec, Françoise Andreu, Marc Rideau, Joël Crèche
ZUSAMMENFASSUNG
In periwinkle cell suspensions, amounts of gibberellic acid ranging from 10 ( - 10) M to 10 ( - 7) M significantly antagonized, in a dose-dependant manner, the stimulation of ajmalicine biosynthesis by cytokinins (CKs). This inhibitory effect was strictly correlated with the abolition of the expression of two genes encoding enzymes of the monoterpenoid indole alkaloid (MIA) biosynthetic pathway and was normally upregulated after CK treatments. Moreover, low concentrations of the gibberellin biosynthesis inhibitor paclobutrazol could reverse the inhibitory effects of low auxin levels on ajmalicine accumulation in the cells. On the other hand, gibberellic acid could not affect the expression of two type-A response regulators considered to be CK primary response genes in periwinkle cells. The antagonistic effects of gibberellins and cytokinins on MIA biosynthesis and their possible impact on elements of the signal transduction are discussed.