- Pregnane glycoside from Hemidesmus indicus as a potential anti-oxidant and anti-dyslipidemic agent.
Pregnane glycoside from Hemidesmus indicus as a potential anti-oxidant and anti-dyslipidemic agent.
Natural product research (2010-02-20)
Arun Sethi, Akriti Bhatia, Sanjay Srivastava, Geetika Bhatia, M M Khan, A K Khanna, J K Saxena
PMID20169502
ZUSAMMENFASSUNG
A new pregnane glycoside hindicusine (1) was isolated from the CHCl(3)-EtOH (3 : 2) extract of Hemidesmus indicus, whose structure was established on the basis of spectroscopic studies. The glycoside (1) and its acetylated derivative (5) were evaluated for their anti-oxidant and anti-dyslipidemic activities.