Merck
  • Synthesis of unnatural selenocystines and beta-aminodiselenides via regioselective ring-opening of sulfamidates using a sequential, one-pot, multistep strategy.
Synthesis of unnatural selenocystines and beta-aminodiselenides via regioselective ring-opening of sulfamidates using a sequential, one-pot, multistep strategy.
The Journal of organic chemistry (2010-04-16)
Nasir Baig Rashid Baig, R N Chandrakala, V Sai Sudhir, Srinivasan Chandrasekaran
ZUSAMMENFASSUNG
A variety of N-alkyl-beta-aminodiselenides have been synthesized in high yield from sulfamidates under mild reaction conditions using potassium selenocyanate and benzyltriethylammonium tetrathiomolybdate ([BnNEt(3)](2)MoS(4)) in a sequential, one-pot, multistep reaction. The tolerance of multifarious protecting groups under the reaction conditions is discussed. The methodology was successfully extended to the synthesis of selenocystine, 3,3'-dialkylselenocystine, and 3,3'-diphenylisoselenocystine and their direct incorporation into peptides.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Selen-L-cystin, 95%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Kaliumselencyanat, reagent grade, 97%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Kaliumselencyanat, ReagentPlus®, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Seleno-DL-cystine, powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.